Learn More
Sino Biological Anti-IL-1 beta Antibody, Rabbit Polyclonal 100 µL
Supplier: Sino Biological 70018T16100
Produced in rabbits immunized with purified, recombinant Canine IL-1 beta ( Catalog#70018-DNAE; Q28292; Ala115-Ser266). IL-1 beta specific IgG was purified by Canine IL-1 beta affinity chromatography.