Learn More
Sino Biological Anti-IL-6R Antibody, Rabbit Polyclonal 100 µL
Supplier:  Sino Biological 50280T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-6R (rM IL-6R; Catalog#50280-M08H; NP_034689.2; Ile44-Glu357). IL-6R specific IgG was purified by Mouse IL-6R affinity chromatography.
Catalog No. 50-007-51427