Learn More
Sino Biological Anti-IL6R (Research Grade Sarilumab Biosimilar) 25 mg
Supplier:  Sino Biological 68072H00125
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Catalog No. 50-007-86496