Learn More
AdipoGen anti-Irisin, pAb (IN102),Polyclonal Antibody
SDP
Supplier:  AdipoGen AG25B0027C100
Encompass_Preferred
anti-Irisin, pAb (IN102), AdipoGen Life Sciences. Polyclonal Antibody. Recognizes human, mouse, rat, monkey Irisin and FNDC5. Detects endogenous mouse and human Irisin [Lit: 1-2]. Source: Rabbit. Applications: WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin activates beige fat cells (beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone Irisin). Irisin is cleaved from the type I membrane protein FNDC5 and improves systemic metabolism by increasing energy expenditure.
Catalog No. 50-463-625