Learn More
Medchemexpress LLC Anti-LILRB4 ILT3 CD8 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9904621MG
Encompass_Preferred
Anti-LILRB4/ILT3/CD85k Antibody is a CHO-expressed humanized antibody that targets LILRB4/ILT3/CD85k The predicted molecular weight (MW) of Anti-LILRB4/ILT3/CD85k Antibody is 144 36 kDa
Catalog No. 50-003-25367