Learn More
Bio X Cell Monoclonal Antibody, Clone AFS98, Mouse, Rat IgG2a
Supplier: Bio X Cell BE0213R005MG
This monoclonal antibody (AFS98 clone) targets mouse colony stimulating factor 1 receptor (CSF1R), also known as macrophage colony-stimulating factor receptor (M-CSFR), and CD115. CSF1R is a single-pass type I membrane protein found on monocytes/macrophages, peritoneal exudate cells, plasmacytoid and conventional dendritic cells, and osteoclasts in mice. It acts as a receptor for CSF1, regulating the proliferation and differentiation of monocytic lineage cells. The AFS98 antibody depletes macrophages and blocks CSFR1 in vivo.
- Clone: AFS98
- Reactivity: mouse
- Isotype: rat IgG2a, κ
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: purified from cell culture supernatant in an animal-free facility
- Purification: protein G
- Molecular weight: 150 kDa
- Conjugation: unconjugated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.