Learn More
Bio X Cell Monoclonal anti-mouse CD3, Clone 17A2, 5 MG
Supplier: Bio X Cell BE0002R005MG
The 17A2 monoclonal antibody reacts with mouse CD3, a transmembrane cell-surface protein that plays crucial roles in TCR signaling, T lymphocyte activation, and antigen recognition. It is expressed on T lymphocytes, NK-T cells, and developing thymocytes.
- Clone: 17A2
- Reactivity: Mouse
- Isotype: Rat IgG2b, κ
- Conjugation: Unconjugated
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 μm filtered
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.