Learn More
Bio X Cell Monoclonal Antibody, anti-mouse CD4, Clone YTS 177, Rat IgG2a
Supplier: Bio X Cell BE00033R100MG
This monoclonal antibody, clone YTS 177, reacts with mouse CD4, a 55 kDa cell surface glycoprotein. CD4 functions as a co-receptor with the T cell receptor, interacting with class II MHC molecules on antigen-presenting cells. It is crucial for T cell development and optimal function. The YTS 177 antibody is non-depleting but induces rapid internalization of CD4, and has been shown to suppress or prevent allograft rejection, allergic reactions, and autoimmune responses.
- Reacts with mouse CD4.
- Rat IgG2a isotype.
- Immunogen: mouse spleen cells.
- Purity ≥95%.
- Sterile (0.2 μm filtration).
- Animal-free production.
- Protein G purification.
- 150 kDa molecular weight.
- Formulation: PBS, pH 7.0, no stabilizers or preservatives.
- Endotoxin ≤1 EU/mg.
- Applications: in vivo blockade of CD4+ T-cell responses, Western blot.
- Store at 4°C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.