Learn More
Bio X Cell Antibody, Anti-Mouse CD47, Clone MIAP301, Rat IgG2a, Unconjugated
Supplier: Bio X Cell BE0270R025MG
The MIAP301 monoclonal antibody specifically targets mouse CD47, an integrin-associated protein (IAP) universally expressed on cells. CD47 is crucial for immune and angiogenic responses, involved in apoptosis, proliferation, adhesion, and migration. This antibody is designed to neutralize CD47 both in vivo and in vitro, valuable for research applications like CD47 blockade and immunofluorescence.
- Reacts with mouse CD47
- Clone MIAP301
- Isotype: Rat IgG2a, κ
- Unconjugated
- Immunogen: Intact CD47 purified from placenta
- Applications: In vivo CD47 blockade, in vitro CD47 blockade, immunofluorescence
- Formulation: PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Produced in an animal-free facility
- Purified using Protein G chromatography
- Molecular weight: 150 kDa
- Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.