Learn More
Bio X Cell InVivoMAb Anti-Mouse CD8 Alpha Antibody, 100 MG
Supplier: Bio X Cell BE0061R100MG
The 2.43 monoclonal antibody reacts with mouse CD8α, a transmembrane glycoprotein co-receptor for the T cell receptor (TCR). CD8α binds to class I MHC molecules on antigen-presenting cells (APC) and is expressed on cytotoxic T cells, thymocytes, natural killer cells, and some dendritic cell subsets. This antibody exhibits depleting activity when used in vivo.
- Isotype: Rat IgG2b, κ
- Conjugation: Unconjugated
- Immunogen: Mouse CTL clone L3
- Reported applications: in vivo CD8+ T cell depletion, Western blot
- Formulation: PBS, pH 7.0
- Endotoxin: ≤1 EU/mg by LAL assay
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 µm filtration
- Production: Animal-free purified
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.