Learn More
Bio X Cell Monoclonal Antibody, mouse CSF1R (CD115), Rat IgG2a, AFS98, Unconjugated
Supplier: Bio X Cell BP0213R050MG
The InVivoPlus anti-mouse CSF1R (CD115) monoclonal antibody, clone AFS98, targets the mouse colony stimulating factor 1 receptor (CSF1R), crucial for monocyte/macrophage proliferation and differentiation. This functional antibody is reported to deplete macrophages and block CSFR1 in vivo. It is supplied as an unconjugated rat IgG2a, kappa antibody, formulated in PBS at pH 7.0.
- In vivo macrophage and monocyte depletion
- In vitro CSF1R neutralization
- Suitable for flow cytometry and Western blot
- Isotype: Rat IgG2a, kappa
- Formulated in PBS, pH 7.0
- Purity: ≥95%
- Endotoxin: ≤0.5 EU/mg
- Aggregation: <5%
- 0.2 μm sterile filtration
- Animal-free production
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.