Learn More
Bio X Cell Antibody, Anti-Mouse CSF1R, Rat IgG2a, 50 MG
Supplier: Bio X Cell BE0213R050MG
The AFS98 monoclonal antibody reacts with mouse colony stimulating factor 1 receptor (CSF1R/CD115), a single-pass type I membrane protein. This antibody has been reported to deplete macrophages and block CSFR1 in vivo.
- Isotype: Rat IgG2a, κ
- Reacts with mouse CSF1R (CD115)
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
- Reported applications: In vivo macrophage depletion, in vitro CSF1R neutralization, in vivo monocyte depletion, flow cytometry, western blot
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.