Learn More
Bio X Cell Monoclonal Antibody, anti-mouse CTLA-4 (CD152), Clone 9D9, Mouse IgG2b
Supplier: Bio X Cell BE0164R050MG
The 9D9 monoclonal antibody targets mouse CTLA-4 (CD152), a 33 kDa cell surface receptor on activated T and B lymphocytes. CTLA-4 negatively regulates cell-mediated immune responses and plays roles in immunological tolerance and protective immunity. As a target for cancer treatment through immune checkpoint blockade, the 9D9 antibody is used in blocking/neutralization experiments and causes intra-tumoral regulatory T cell depletion.
- Host: Mouse
- Isotype: Mouse IgG2b
- Reacts with: Mouse
- Formulation: PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95%
- Sterility: 0.2 µm filtration
- Applications: In vivo CTLA-4 neutralization, western blot, in vivo intra-tumoral regulatory T cell depletion, in vitro organoids/organ-on-chip
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.