Learn More
Bio X Cell InVivoMAb Antibody, Anti-Mouse IFNγR, Rat IgG2a, Clone GR-20
Supplier: Bio X Cell BE0029R005MG
The GR-20 monoclonal antibody interacts with the mouse IFNγR (interferon gamma receptor) α chain (CD119). CD119 forms a heterodimer to create the IFNγR, a Class II cytokine receptor found on nearly all cell types. The GR-20 antibody targets an epitope within the receptor's ligand-binding site, preventing IFNγ from binding and inhibiting IFNγ-mediated effects.
- Clone GR-20
- Isotype rat IgG2a, κ
- Reacts with mouse IFNγR (CD119)
- Immunogen: BALB/c mouse monomyelocytic cell line WEHI-3
- Applications: In vivo and in vitro IFNγR neutralization
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtered
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.