Learn More
Bio X Cell Monoclonal Antibody, anti-mouse IL-10R (CD210), Clone 1B1.3A
Supplier: Bio X Cell BE0050R100MG
The 1B1.3A monoclonal antibody targets mouse IL-10R (IL-10 receptor), also known as CD210. This class II cytokine receptor is found on various cell types including thymocytes, T lymphocytes, B lymphocytes, NK cells, monocytes, and macrophages. This neutralizing antibody blocks human IL-10 binding, which cross-reacts with mouse IL-10R, but does not recognize human IL-10R.
- Clone 1B1.3A
- Isotype rat IgG1, κ
- Reacts with mouse
- Blocks IL-10/IL-10R signaling in vivo and in vitro
- Used in flow cytometry and Western blot
- Unconjugated format
- Immunogen: recombinant ligand-binding domain of mouse IL-10R
- Formulation: PBS, pH 6.5, 0.01% Tween; no stabilizers or preservatives
- Endotoxin: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtered
- Production: animal-free facility, protein G purified
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.