Learn More
Bio X Cell InVivoMAb Antibody, anti-mouse IL-2, Clone JES6-1A12
Supplier:  Bio X Cell BE0043R005MG
Encompass_Preferred
The JES6-1A12 monoclonal antibody reacts with mouse IL-2, a 17 kDa cytokine primarily produced by T cells in response to antigenic or mitogenic stimulation. IL-2 is essential for T cell proliferation and other activities crucial for regulating immunity. It can also stimulate the growth and differentiation of B cells, monocytes/macrophages, and NK cells. Additionally, IL-2 prevents autoimmune diseases by promoting the differentiation of certain immature T cells into regulatory T cells. This antibody has been shown to neutralize IL-2 in vivo.
  • Neutralizes IL-2 in vivo.
  • Stimulates IL-2 receptor in vivo as a complex with IL-2.
  • Purified from cell culture supernatant in an animal-free facility.
  • Filtered to 0.2 μM for sterility.
Catalog No. 50-139-2821