Learn More
Bio X Cell Monoclonal Antibody, Clone 1A8, Mouse Ly6G, Rat IgG2a
Supplier: Bio X Cell BE00751R005MG
The 1A8 monoclonal antibody reacts with mouse Ly6G, a GPI-anchored cell surface protein found on myeloid lineage cells, including granulocytes and peripheral neutrophils. This antibody specifically targets Ly6G without cross-reactivity to Ly6C, making it a valuable tool for identifying these cell populations.
- Specifically reacts with mouse Ly6G with no cross-reactivity to Ly6C.
- Suitable for in vivo neutrophil and MDSC depletion.
- Validated for immunofluorescence, immunohistochemistry (paraffin and frozen), and flow cytometry.
- Formulated in PBS, pH 7.0, without stabilizers or preservatives.
- Purity of 95% or greater and endotoxin level of 1 EU/mg or less.
- Isotype: rat IgG2a, κ.
- Purified by protein G.
- Store at 4°C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.