Learn More
Bio X Cell Antibody, anti-mouse Ly6G/Ly6C (Gr-1), Rat, Monoclonal, RB6-8C5
Supplier: Bio X Cell BP0075R005MG
The Bio X Cell InVivoPlus anti-mouse Ly6G/Ly6C (Gr-1) monoclonal antibody, clone RB6-8C5, is a rat-derived antibody. It strongly reacts with mouse Ly6G and weakly with Ly6C (previously GR-1), both GPI-anchored cell surface proteins of the myeloid lineage. This antibody effectively marks mature granulocytes and peripheral neutrophils.
- Host: Rat, clone: RB6-8C5, isotype: Rat IgG2b, κ.
- Immunogen: Mouse granulocytes.
- Applications: In vivo depletion of Gr-1+ myeloid cells, flow cytometry, immunohistochemistry (paraffin and frozen).
- Formulation: PBS, pH 7.0, no stabilizers or preservatives.
- Quality control: Endotoxin ≤0.5 EU/mg, aggregation <5%, purity ≥95%.
- Production: Sterility by 0.2 μm filtration, purified using Protein G.
- Molecular weight: 150 kDa.
- Storage: 4°C, do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.