Learn More
Bio X Cell Antibody, M5/114 Clone, Mouse Reactivity, Rat IgG2b Isotype, 5 mg
Supplier: Bio X Cell BE0108R005MG
This monoclonal antibody reacts with specific mouse MHC Class II haplotypes, including I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, while demonstrating no reactivity with I-Af, I-Ak, or I-As haplotypes. It is reported to inhibit I-A-restricted T cell responses.
- Reacts with mouse MHC Class II haplotypes I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek
- Inhibits I-A-restricted T cell responses
- Isotype: Rat IgG2b
- Purity: ≥95% by SDS-PAGE
- Formulated in PBS, pH 7.0, with no stabilizers or preservatives
- Produced in an animal-free facility
- Suitable for in vivo MHC II blockade, functional assays, immunofluorescence, western blot, immunoprecipitation, and flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.