Learn More
Bio X Cell Monoclonal Antibody, anti-mouse PD-L1 10F.9G2, 50 MG
Supplier: Bio X Cell BE0101R050MG
The 10F.9G2™ monoclonal antibody reacts with mouse PD-L1 (programmed death ligand 1), also known as B7-H1 or CD274. It blocks the interaction between PD-L1 and PD-1, and between PD-L1 and B7-1 (CD80), playing a role in tumor immune evasion research.
- Purified via protein G
- Formulated in PBS, pH 6.5, without stabilizers or preservatives
- Endotoxin ≤1 EU/mg
- Purity ≥95% by SDS-PAGE
- Sterility by 0.2 μm filtration
- Suitable for in vivo and in vitro PD-L1 blockade
- Validated for immunofluorescence, immunohistochemistry (frozen), flow cytometry, and Western blot
- Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.