Learn More
Bio X Cell Antibody, anti-mouse PD-L1 (B7-H1), Clone 10F.9G2
Supplier: Bio X Cell BP0101R025MG
This monoclonal antibody, clone 10F.9G2, targets mouse PD-L1 (B7-H1/CD274), a key protein in tumor immune evasion. It blocks PD-L1 interaction with PD-1 and B7-1 (CD80), inhibiting T cell proliferation. Supplied in PBS, pH 6.5, without stabilizers, it is suitable for various research applications.
- Monoclonal antibody, clone 10F.9G2
- Reacts with mouse PD-L1 (B7-H1/CD274)
- Host species: rat; Isotype: IgG2b, κ
- Molecular weight: 150 kDa
- Supports in vivo and in vitro PD-L1 blockade
- Validated for immunofluorescence, IHC (frozen), flow cytometry, and Western blot
- Formulated in PBS, pH 6.5, no stabilizers or preservatives
- Low endotoxin levels (<1 EU/mg)
- High purity (≥95%) and low aggregation (<5%)
- Sterile (0.2 µm filtered)
- Protein G purified from an animal-free facility
- Store at 4°C; do not freeze; one-year shelf-life
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.