Learn More
Bio X Cell InVivoMab anti-mouse TIGIT, Clone: 1G9, Size: 50 MG
Supplier:  Bio X Cell BE0274R050MG
Encompass_Preferred
InVivoMab anti-mouse TIGIT, Clone: 1G9, Size: 50 MG, Isotype: mouse IgG1, kappa, Applications: in vivo TIGIT blockade, in vitro TIGIT blockade, Flow cytometry
Catalog No. 50-139-3390