Learn More
Bio X Cell Antibody, Mouse TIGIT, Mouse IgG1, Unconjugated, Protein G Purified
Supplier: Bio X Cell BE0274R005MG
The 1G9 monoclonal antibody targets mouse TIGIT (T cell immunoreceptor with Ig and ITIM domains), a 26 kDa type I transmembrane protein. TIGIT, expressed on follicular T helper cells in mice and various T cell subsets and NK cells in humans, negatively regulates NK and T cell activation. Its binding on T cells by dendritic cells promotes a tolerogenic phenotype.
- Isotype: mouse IgG1, κ
- Recommended isotype control: mouse IgG1 isotype control
- Recommended dilution buffer: pH 7.0 dilution buffer
- Conjugation: unconjugated
- Immunogen: mouse TIGIT
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1 EU/mg
- Purity: ≥95% (determined by SDS-PAGE)
- Sterility: 0.2 μm filtration
- Purification: protein G
- Molecular weight: 150 kDa
- Storage: store at 4°C, do not freeze
- Applications: in vivo TIGIT stimulation, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.