Learn More
Bio X Cell Antibody, Clone XT3.11, Rat IgG1, Unconjugated
Supplier: Bio X Cell BE0058R005MG
The XT3.11 monoclonal antibody reacts with mouse TNFα (tumor necrosis factor-alpha), a multifunctional proinflammatory cytokine. It plays key roles in immunoregulation, cell proliferation, differentiation, apoptosis, antitumor activity, and inflammation. This antibody is suitable for in vivo and in vitro TNFα neutralization, and Western blot applications.
- Reacts with mouse TNFα
- Isotype: Rat IgG1
- Suitable for in vivo TNFα neutralization
- Suitable for in vitro TNFα neutralization
- Suitable for Western blot applications
- Immunogen: Recombinant mouse TNFα
- Formulated in PBS, pH 8.0, with no stabilizers or preservatives
- Endotoxin: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Purified from cell culture supernatant in an animal-free facility
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.