Learn More
BIOHIPPO Anti-MAT2A Polyclonal Antibody, 100 µg
SDP
Supplier:  BIOHIPPO BHA21404275
Encompass_Preferred
Human MAT2A Polyclonal Antibody is a Rabbit-derived polyclonal antibody that specifically recognizes MAT2A. It shows reactivity with Human, Mouse, Rat. Validated applications include ELISA, IHC, WB. Purified by antigen affinity column. Supplied as 100 ug.
Catalog No. 50-008-60602