Learn More
Medchemexpress LLC Anti-Mouse CD20 Anti 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP9901395MG
Encompass_Preferred
Anti-Mouse CD20 Antibody (AISB12) is an anti-mouse CD20 IgG2a monoclonal antibody Anti-Mouse CD20 Antibody (AISB12) can deplete B cells Anti-Mouse CD20 Antibody (AISB12) can be used for research on immunology[1][2]
Catalog No. 50-003-24658