Learn More
Medchemexpress LLC Anti-Mouse E-selecti 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9902471MG
Encompass_Preferred
Anti-Mouse E-selectin/CD62E Antibody (9A9) is an anti-mouse E-selectin/CD62E IgG2b monoclonal antibody Anti-Mouse E-selectin/CD62E Antibody (9A9) has an inhibitory effect on cancer cell colonization Anti-Mouse E-selectin/CD62E Antibody (9A9) can be used for researches on inflammation conditions and cancer such as acute microvascular inflammation and breast cancer[1][2]
Catalog No. 50-003-25392