Learn More
Medchemexpress LLC Anti-Mouse Galectin- 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP99024210MG
Encompass_Preferred
Anti-Mouse Galectin-9 Antibody (RG9-1) is an anti-mouse Galectin-9 IgG2b monoclonal antibody Anti-Mouse Galectin-9 Antibody (RG9-1) promotes inflammatory response by enhancing the secretion of IFN- TNF- and IL-6 Anti-Mouse Galectin-9 Antibody (RG9-1) can increase CD8 T cell and Treg frequency Anti-Mouse Galectin-9 Antibody (RG9-1) reduces graft survival time by increasing CD4 and CD8 T cell infiltration Anti-Mouse Galectin-9 Antibody (RG9-1) can be used for researches on inflammation conditions cancer and xenotransplantation such as ischemia-reperfusion colon cancer breast cancer and leukemia[1][2][3][4]
Catalog No. 50-003-25350