Learn More
Medchemexpress LLC Anti-Mouse Human Mac 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP9901985MG
Encompass_Preferred
Anti-Mouse/Human Mac-2/Galectin-3 Antibody (TIB-166) is a rat-derived anti-mouse/human Mac-2/Galectin-3 IgG2a monoclonal antibody Anti-Mouse/Human Mac-2/Galectin-3 Antibody (TIB-166) significantly inhibits IL-6 expression (inhibition rates 34 7% and 55 3%) Anti-Mouse/Human Mac-2/Galectin-3 Antibody (TIB-166) is often used for immunoprecipitation western blot immunofluorescence and flow cytometry[1][2][3][4]
Catalog No. 50-003-24864