Learn More
Medchemexpress LLC Anti-Mouse Human pho 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP99016910MG
Encompass_Preferred
Anti-Mouse/Human phosphorylated PD-1/CD279 Antibody (407 6G12) is a mouse-derived IgG2a type antibody inhibitor targeting to mouse/human phosphorylated PD-1/CD279 Anti-Mouse/Human phosphorylated PD-1/CD279 Antibody (407 6G12) can detect the phosphorylated form of the PD-1 ITSM by both western blot and flow cytometry[1]
Catalog No. 50-003-25285