Learn More
Selleck Chemical LLC Anti-mouse/human VLA-4 -InVivo
SDP
Supplier:  Selleck Chemical LLC A21625MG 20PC
Encompass_Preferred
Anti-mouse/human VLA-4 -InVivo reacts with human and mouse VLA-4 chain also known as CD49d and integrin alpha 4 VLA-4 is a 150 kDa glycoprotein belonging to the integrin family that is expressed by many cell types including T and B lymphocytes monocytes eosinophils basophils mast cells thymocytes NK cells and dendritic cells
Catalog No. 50-313-8068