Learn More
Medchemexpress LLC Anti-Mouse IFN gamma 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP99079810MG
Encompass_Preferred
Anti-Mouse IFN gamma Antibody (XMG1 2) is a kind of rat-derived IgG1 antibody inhibitor targeting to IFN- Anti-Mouse IFN gamma Antibody (XMG1 2) can neutralize IFN- Anti-Mouse IFN gamma Antibody (XMG1 2) can be used for the researches of cancer inflammation and immunology such as multiple myeloma and chronic inflammation[1][2][3][4][5]
Catalog No. 50-003-24151