Learn More
REVMAB BIOSCIENCES USA INC IgG2b Rabbit Anti-Mouse Monoclonal Antibody [RM108]
Supplier:  REVMAB BIOSCIENCES USA INC 31100400
Encompass_Preferred
Anti-Mouse IgG2b rabbit monoclonal antibody, Clone RM108 (100UG) ; WB (stronger on nonreduced), IP, ICC, IHC, FC, ELISA validated; Product info: www.revmab.com
Catalog No. NC1729082