Learn More
ECM Bioscience Anti-Mouse Ig:HRP Donkey Polyclonal, ECM Biosciences
Supplier:  ECM Bioscience MS3001
Encompass_Preferred
This antibody has been pre-adsorbed with various immunoglobulins from non-mouse species before affinity-purification using mouse IgG coupled to agarose beads. Purified donkey polyclonal antibody was conjugated to horseradish peroxidase (HRP). This secondary reagent can be used to detect mouse immunoglobulins in various antibody applications that utilize HRP readouts, such as western blot, ELISA, immunocytochemistry, and immunohistochemistry. The immunogen was The HRP-conjugated donkey polyclonal secondary reagent reacts with the whole molecule mouse IgG, and the light chains of other mouse immunoglobulins. The secondary reagent has minimal cross-reactivity with bovine, chicken, goat, horse, human, rabbit, rat, and sheep immunoglobulins. Accession #. Applications = WB, E, ICC, IHC; Species Reactivity = Ms; Size = 100 μl; Buffer = Donkey polyclonal antibody is supplied in 100µl PBS, 50% glycerol, and 1 mg/ml BSA.
Catalog No. 50-206-8757