Learn More
Medchemexpress LLC Anti-Mouse IL-15 Ant 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9902231MG
Encompass_Preferred
Anti-Mouse IL-15 Antibody (AIO 3) is a rat-derived IgG2a type antibody inhibitor targeting to mouse IL-15 Anti-Mouse IL-15 Antibody (AIO 3) can neutralize IL-15 Anti-Mouse IL-15 Antibody (AIO 3) can be used for the researches of cancer infection and inflammation such as hepatitis virus infection and esophagitis[1][2][3][4][5]
Catalog No. 50-003-25190