Learn More
Medchemexpress LLC Anti-Mouse IL-18 Ant 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP9902205MG
Encompass_Preferred
Anti-Mouse IL-18 Antibody (YIGIF74-1G7) is a rat-derived IgG2a type antibody inhibitor targeting to mouse IL-18 Anti-Mouse IL-18 Antibody (YIGIF74-1G7) can neutralize IL-18 Anti-Mouse IL-18 Antibody (YIGIF74-1G7) can be used for the researches of cancer infection inflammation and immunology such as hepatocellular carcinoma and eosinophilic esophagitis[1][2][3]
Catalog No. 50-003-25805