Learn More
Medchemexpress LLC Anti-Mouse IL-27 p28 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP9902155MG
Encompass_Preferred
Anti-Mouse IL-27 p28 Antibody (MM27 7B1) is a mouse-derived anti-mouse IL-27 p28 IgG2a type antibody inhibitor Anti-Mouse IL-27 p28 Antibody (MM27 7B1) can neutralize IL-27 Anti-Mouse IL-27 p28 Antibody (MM27 7B1) delays CD8 T cell reconstitution mediated by IL-27 after murine anti-thymocyte globulin (mATG) treatment in BALB/c heart allografts mice models[1][2][3]
Catalog No. 50-003-25801