Learn More
Medchemexpress LLC Anti-Mouse IL-9 Anti 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP99020810MG
Encompass_Preferred
Anti-Mouse IL-9 Antibody (9C1) is an anti-mouse IL-9 IgG2a monoclonal antibody Anti-Mouse IL-9 Antibody (9C1) can reduce the activation of NF- B signaling pathway and decrease the infiltration of inflammatory cells Anti-Mouse IL-9 Antibody (9C1) effectively reduces Th9 cell-mediated allergic reactions and anti-tumor effects Anti-Mouse IL-9 Antibody (9C1) can be used for researches on inflammation infection conditions and cancer such as parasitic infections allergic reactions breast cancer and osteosarcoma[1][2][3][4]
Catalog No. 50-003-25756