Learn More
Biolegend Brilliant Violet 605™ anti-mouse Ly-6G
Supplier:  Biolegend 127639
Encompass_Preferred
Brilliant Violet 605™ anti-mouse Ly-6G [1A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1074525