Learn More
Medchemexpress LLC Anti-Mouse Ly6G Anti 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9901181MG
Encompass_Preferred
Anti-Mouse Ly6G Antibody (1A8) is a rat-derived anti-mouse Ly6G IgG2a type antibody inhibitor Anti-Mouse Ly6G Antibody (1A8) can deplete neutrophil in various mice Anti-Mouse Ly6G Antibody (1A8) can be used for the researches of inflammation such as arthritis and peritonitis[1][2]
Catalog No. 50-003-24681