Learn More
Bio X Cell InVivoMab anti-mouse OX40L (CD134L), Clone: RM134L, Size: 100 mg
Supplier:  Bio X Cell BE00331R100MG
Encompass_Preferred
InVivoMab anti-mouse OX40L (CD134L), Clone: RM134L, Size: 100 mg, Isotype: rat IgG2b, κ, Applications: in vivo blocking of OX40/OX40L signaling, in vitro OX40L neutralization
Catalog No. NC1977787