Learn More
Selleck Chemical LLC Anti-mouse PD-L1-InVivo
SDP
Supplier:  Selleck Chemical LLC A21155MG 5PC
Encompass_Preferred
Anti-mouse PD-L1-InVivo (Clone 10F 9G2) monoclonal antibody reacts with mouse PD-L1 (also known as B7-H1 CD274) The 10F 9G2 antibody has been shown to block the interaction between PD-L1 and PD-1 and between PD-L1 and B7-1 (CD80)
Catalog No. 50-313-8018