Learn More
Medchemexpress LLC Anti-Mouse Rat Human 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9901171MG
Encompass_Preferred
Anti-Mouse/Rat/Human Osteopontin/SPP1 Antibody (MPIIIB10) is a mouse-derived Osteopontin/SPP1 IgG1 type antibody inhibitor Anti-Mouse/Rat/Human Osteopontin/SPP1 Antibody (MPIIIB10) blocks Angiotensin II (HY-13948)-induced DNA synthesis and collagen gel contraction in cardiac fibroblasts Anti-Mouse/Rat/Human Osteopontin/SPP1 Antibody (MPIIIB10) significantly inhibits tumor growth in CT26 or MC38 tumors mice models[1][2][3]
Catalog No. 50-003-24386