Learn More
Medchemexpress LLC Anti-Mouse TIM-4 Ant 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9908201MG
Encompass_Preferred
Anti-Mouse TIM-4 Antibody (RMT4-53) is a rat-derived IgG2b type antibody inhibitor targeting to mouse TIM-4 Anti-Mouse TIM-4 Antibody (RMT4-53) can block T cell immunoglobulin and mucin domain 4 (TIM-4) Anti-Mouse TIM-4 Antibody (RMT4-53) can inhibit the engulfment of apoptotic cells and T cell proliferation Anti-Mouse TIM-4 Antibody (RMT4-53) can be used for the researches of cancer immunology and metabolic disease such as MC38 tumor and graft[1][2][3]
Catalog No. 50-003-23473