Learn More
Medchemexpress LLC Anti-Mouse VEGF-A An 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP9901485MG
Encompass_Preferred
Anti-Mouse VEGF-A Antibody (2G11-2A05) is a rat-derived IgG2a type antibody inhibitor tragrting to mouse VEGF-A with high affinity Anti-Mouse VEGF-A Antibody (2G11-2A05) shows good anti-tumor effect in gastric cancer xenograft models[1]
Catalog No. 50-003-25974