Learn More
Creative Biolabs Anti-Mouse CXCL13 Recombinant scFv Fragment, 2C4, 1 mg, Unconjugated, High affinity, ELISA application

Supplier: Creative Biolabs TAB112MZ1MG
This technical summary describes the Anti-Mouse CXCL13 Recombinant Antibody, clone 2C4, supplied by Creative Biolabs. This recombinant single-chain variable fragment (scFv) antibody is engineered for high affinity and specificity to mouse CXCL13. It is a valuable tool for scientific research, particularly for the detection and inhibition of mouse CXCL13 activity. Its inhibitory capabilities make it potentially useful in studies related to disorders where CXCL13 activity is detrimental.
- Target: Mouse CXCL13
- Clone: 2C4
- Antibody type: Recombinant scFv fragment
- Reactivity: Mouse
- Affinity: High
- Application: ELISA
- Function: Detection of mouse CXCL13
- Function: Inhibition of mouse CXCL13 activity
- Quantity: 1 mg
- Conjugation: Unconjugated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.