Learn More
Seracare Life Sciences Inc ANTI-MSIGG(G)F(AB )2HSA DL488
Supplier:  Seracare Life Sciences Inc 52300431
Encompass_Preferred
Affinity purified polyclonal antibody fragment, F(ab')2, to Mouse IgG heavy chains (gamma), made in Goat and labeled with DyLight 488. Cross-reactivity with human serum has been minimized with affinity procedures. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-673-26