Learn More
Cusabio Technology LLC Human anti-Mullerian hormone (AMH) ELISA kit
Supplier:  Cusabio Technology LLC CSBE12756H
Encompass_Preferred
This Human AMH ELISA Kit was designed for the quantitative measurement of Human AMH protein in serum, plasma, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 62.5 pg/mL-4000 pg/mL and the sensitivity is 15.6 pg/mL.
Catalog No. NC3654179