Learn More
Selleck Chemical LLC Anti-Myc magnetic beads
SDP
Supplier:  Selleck Chemical LLC B263025 ML
Encompass_Preferred
Anti-Myc magnetic beads kit is based on hydroxyl magnetic beads covalently coupling with mouse IgG1 monoclonal antibody it is recommended to use for co-immunoprecipitation and protein purification
Catalog No. 50-313-5685