Learn More
Advanced Targeting Systems NGFr (ME20.4, p75) Mouse Monoclonal, 50mcg
Supplier:  Advanced Targeting Systems ABN07
Encompass
The p75 neurotrophin receptor (p75NTR), also known as the low affinity nerve growth factor receptor, binds nerve growth factor, brain-derived neurotrophic factor, neurotrophin-3 and neurotrophin-4 with varying specificities. The p75NTR plays an important role in neurotrophic factor signaling and has been shown to modulate the susceptibility of selective cellular populations to programmed cell death. This antibody recognizes the p75NTR (low affinity neurotrophin receptor) in human, primate, rabbit, sheep, dog, cat, and pig. It was derived from immunization of mice with WM245 melanoma cells. The antibody is routinely tested by flow cytometry. Applications include flow cytometry, immunoprecipitation, immunohistochemistry, electron microscopy, immunocytochemistry, radioimmunoassay, and targeting.
Catalog No. NC0468344